Learn More
Waters Corp Waters Sentry Guard Cartridge, Shield RP18, 3.5 μm, 4.6 mm X 20 mm
Supplier:  Waters Corp 186003070
Encompass_Preferred
These Sentry Guard Cartridges complement C18 and C8 phases, offering a powerful tool for HPLC method development. Their embedded carbamate group in the bonded phase ligand provides alternate selectivity, particularly for phenolic compounds.
  • Extends HPLC column life by minimizing particle matter and chemical pollutants.
  • Dual holder designs allow tool-free replacement with integrated or any HPLC column.
  • Carbamate group provides alternate selectivity for phenolic compounds.
  • Shield technology reduces silanol interactions, achieving symmetrical peaks and enhanced peak shape.
  • Maximizes particle efficiency, delivering unrivaled peak shape and capacity.
  • Offers extended column lifetime and assay robustness through improved pH stability.
Catalog No. 50-781-522