Learn More
Waters Corp XBridge BEH Shield RP18 Sentry Guard Cartridge, 130Å, 5 µm, 4.6 mm X 20 mm, 2/pk
Catalog No. 50781525 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
Catalog No. 50-781-525 Supplier Waters Corp Supplier No. 186003073
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.