Learn More
Waters Corp Waters XBridge BEH Shield RP18 IS Column, 130Å, 5 μm, 2.1 mm × 30 mm
Catalog No. 50781589 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 columns are designed to complement XBridge BEH C18 and C8 phases, serving as a significant tool for HPLC method development. The column features an embedded carbamate group within its bonded phase ligand, which offers an alternative selectivity, particularly for phenolic compounds, when compared to traditional straight chain alkyl columns.
  • Provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns, due to the embedded carbamate group in the bonded phase ligand.
  • Complements BEH C18 and C8 phases.
  • Powerful tool for HPLC method development.
Catalog No. 50-781-589 Supplier Waters Corp Supplier No. 186003157
XBridge BEH Shield RP18 columns are designed to complement XBridge BEH C18 and C8 phases, serving as a significant tool for HPLC method development. The column features an embedded carbamate group within its bonded phase ligand, which offers an alternative selectivity, particularly for phenolic compounds, when compared to traditional straight chain alkyl columns.
  • Provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns, due to the embedded carbamate group in the bonded phase ligand.
  • Complements BEH C18 and C8 phases.
  • Powerful tool for HPLC method development.