Learn More
Waters Corp XBridge BEH Shield RP18 Method Validation Kit, 130Å, 3.5 µm, 3 mm X 100 mm, 3/pk
Catalog No. 50465970 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
Catalog No. 50-465-970 Supplier Waters Corp Supplier No. 186003789
Add to Cart
Add to Cart
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.