Learn More
Waters Corp Waters XBridge BEH Shield RP18 OBD Prep Column, 19 x 100 mm, 5 μm
Supplier:  Waters Corp 186002986
Encompass_Preferred
This XBridge preparative column features BEH Shield RP18 chemistry and is designed for high-performance liquid chromatography. It has dimensions of 19 x 100 mm and a particle size of 5 μm.
  • Embedded Polar C18 provides different selectivity.
  • Improved peak shape for basic compounds.
Catalog No. 50-781-442