Learn More
Waters Corp Waters OBD Prep Column, BEH Shield RP18, 5 μm, 130 Å, 30 mm × 150 mm
Catalog No. 50781446 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 columns are essential tools for HPLC method development, offering alternate selectivity particularly for phenolic compounds. An embedded carbamate group improves peak shape for basic compounds. Utilizing BEH particle technology, these columns are stable across a wide pH range and at high temperatures.
  • Alternate selectivity, beneficial for phenolic compounds.
  • Improved peak shape for basic compounds with embedded carbamate group.
  • BEH technology for universal compatibility and excellent peak shape.
  • Stable across a pH range of 2 to 11.
  • Stable at temperatures up to 50°C (low pH) and 45°C (high pH).
  • Maximum pressure of 6000 psi (415 Bar).
  • Spherical particles, 5 μm size, 130 Å pore size.
  • Compatible with LC/MS and LC techniques.
Catalog No. 50-781-446 Supplier Waters Corp Supplier No. 186002990
Add to Cart
Add to Cart
Request a Quote
XBridge BEH Shield RP18 columns are essential tools for HPLC method development, offering alternate selectivity particularly for phenolic compounds. An embedded carbamate group improves peak shape for basic compounds. Utilizing BEH particle technology, these columns are stable across a wide pH range and at high temperatures.
  • Alternate selectivity, beneficial for phenolic compounds.
  • Improved peak shape for basic compounds with embedded carbamate group.
  • BEH technology for universal compatibility and excellent peak shape.
  • Stable across a pH range of 2 to 11.
  • Stable at temperatures up to 50°C (low pH) and 45°C (high pH).
  • Maximum pressure of 6000 psi (415 Bar).
  • Spherical particles, 5 μm size, 130 Å pore size.
  • Compatible with LC/MS and LC techniques.