Learn More
Waters Corp Waters XBridge Preparative Column, BEH Shield RP18, 5 µm, 30 x 100 mm
Catalog No. 50781445 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge Preparative Column is an OBD type column designed for preparative chromatography. It features BEH Shield RP18 chemistry, a particle size of 5 µm, a pore diameter of 130 Å, and dimensions of 30 x 100 mm.
  • OBD column type for preparative applications.
  • BEH Shield RP18 chemistry provides unique selectivity.
  • 5 µm particle size enables efficient separations.
  • 130 Å pore diameter for optimized analyte interaction.
  • Dimensions of 30 x 100 mm for specific chromatography requirements.
Catalog No. 50-781-445 Supplier Waters Corp Supplier No. 186002989
Add to Cart
Add to Cart
Request a Quote
This XBridge Preparative Column is an OBD type column designed for preparative chromatography. It features BEH Shield RP18 chemistry, a particle size of 5 µm, a pore diameter of 130 Å, and dimensions of 30 x 100 mm.
  • OBD column type for preparative applications.
  • BEH Shield RP18 chemistry provides unique selectivity.
  • 5 µm particle size enables efficient separations.
  • 130 Å pore diameter for optimized analyte interaction.
  • Dimensions of 30 x 100 mm for specific chromatography requirements.