Learn More
Waters Corp Waters Corporation OBD Prep Column, BEH Shield RP18, 5 μm, 19 mm x 150 mm
Catalog No. 50781443 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XBridge BEH Shield RP18 OBD Prep Columns are designed for HPLC method development and preparative liquid chromatography, offering unique selectivity and high performance.
  • Provides alternate selectivity for phenolic compounds via an embedded carbamate group.
  • Features reversed phase separation with hybrid, spherical BEH particles (5 μm, 130 Å pore).
  • Stable across a pH range of 2 to 11 (max 50 °C low pH / 45 °C high pH), up to 6000 psi (415 Bar).
  • Low silanol activity with Shield RP18 bonding and TMS endcap style.
  • 19 mm inner diameter by 150 mm length OBD preparative column format.
  • Uniform packed bed density ensures direct scalability from analytical to preparative methods.
  • Excellent column lifetime and consistent performance deliver optimal resolution, peak shape, and mass loading.
Catalog No. 50-781-443 Supplier Waters Corp Supplier No. 186002987
These XBridge BEH Shield RP18 OBD Prep Columns are designed for HPLC method development and preparative liquid chromatography, offering unique selectivity and high performance.
  • Provides alternate selectivity for phenolic compounds via an embedded carbamate group.
  • Features reversed phase separation with hybrid, spherical BEH particles (5 μm, 130 Å pore).
  • Stable across a pH range of 2 to 11 (max 50 °C low pH / 45 °C high pH), up to 6000 psi (415 Bar).
  • Low silanol activity with Shield RP18 bonding and TMS endcap style.
  • 19 mm inner diameter by 150 mm length OBD preparative column format.
  • Uniform packed bed density ensures direct scalability from analytical to preparative methods.
  • Excellent column lifetime and consistent performance deliver optimal resolution, peak shape, and mass loading.