Learn More
Waters Corp XBridge Oligonucleotide BEH C18 OBD Prep Column, 130Å, 2.5 µm, 10 mm X 50 mm, 1/pk
Supplier:  Waters Corp 186008212
Encompass_Preferred
Waters Oligonucleotide Separation Technology columns are based on the second generation of hybrid-silica BEH Technology particles functionalized with C18. The separation of detritylated synthetic oligonucleotide samples is based on the well-established method of ion-pair, reversed-phase chromatography. The availability of 1.7µm UPLC technology or 2.5µm HPLC particles in various column dimensions provides flexibility to meet various laboratory-scale isolation or analysis needs and delivers exceptional sample resolution and superior column life. In addition, Waters manufacturing and quality control testing procedures help ensure consistent batch-to-batch and column-to-column performance regardless of application demands.
Catalog No. 50-138-4699