Learn More
Waters Corp Waters XBridge OBD Prep Column, C8, 5 μm, 130 Å, 10x50 mm
Catalog No. 501384659 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 OBD Prep Column is designed for efficient preparative chromatography, offering enhanced performance with its trifunctionally bonded C8 sorbent. It provides less retention than C18 columns and maintains stability across a wide pH range (1-12), ensuring excellent peak shape and consistent results for isolating target compounds.
  • Lower retention than C18 sorbents.
  • Accelerates method development.
  • Delivers excellent peak shape and chromatographic performance.
  • Stable across a wide pH range (1-12).
  • Features superior low pH stability and ultra-low bleed.
  • Utilizes OBD Technology for uniform packed bed density.
  • Offers direct scalability from analytical to preparative.
  • Ensures long column lifetime and consistent performance.
  • Maximizes recovery with robust packing.
Catalog No. 50-138-4659 Supplier Waters Corp Supplier No. 186008172
The XBridge BEH C8 OBD Prep Column is designed for efficient preparative chromatography, offering enhanced performance with its trifunctionally bonded C8 sorbent. It provides less retention than C18 columns and maintains stability across a wide pH range (1-12), ensuring excellent peak shape and consistent results for isolating target compounds.
  • Lower retention than C18 sorbents.
  • Accelerates method development.
  • Delivers excellent peak shape and chromatographic performance.
  • Stable across a wide pH range (1-12).
  • Features superior low pH stability and ultra-low bleed.
  • Utilizes OBD Technology for uniform packed bed density.
  • Offers direct scalability from analytical to preparative.
  • Ensures long column lifetime and consistent performance.
  • Maximizes recovery with robust packing.