Learn More
Waters Corp Waters XBridge Protein BEH C4 Column, 300Å, 2.5 μm
Supplier:  Waters Corp 186009138
Encompass_Preferred
The XBridge Protein BEH C4 Column is optimized and QC tested with the MassPREP Protein Mix standard for protein separations. The 300Å pore size trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for various protein-based applications.
  • Designed and quality control (QC) tested for protein separations.
  • Features a 2.5 μm BEH particle with 300Å pore size.
  • Offers the widest usable pH range, superior low pH stability, and ultra-low column bleed.
  • Suitable for low-level intact protein and subunit analysis.
Catalog No. NC1604316