Learn More
MVE BIOLOGICAL SOLUTIONS MVE XC 47/11-6 Cryogenic Freezer, 47.4 L LN2 Capacity, 1,320 Vial Capacity, 120.4 lbs Full Weight
Catalog No. NC1421320
Encompass_Preferred
The MVE XC 47/11-6 Cryogenic Freezer is a reliable solution for large-capacity cryogenic storage, maximizing storage efficiency and minimizing nitrogen loss. Its lightweight aluminum construction and advanced vacuum retention system ensure optimal temperatures for biological samples. With an LN2 capacity of 47.4 L, it supports the organization of up to 1,320 vials or 6,216 straws, making it an essential unit in scientific environments demanding durability and performance.
Catalog No. NC1421320 Supplier MVE BIOLOGICAL SOLUTIONS Supplier No. 10719924
The MVE XC 47/11-6 Cryogenic Freezer is a reliable solution for large-capacity cryogenic storage, maximizing storage efficiency and minimizing nitrogen loss. Its lightweight aluminum construction and advanced vacuum retention system ensure optimal temperatures for biological samples. With an LN2 capacity of 47.4 L, it supports the organization of up to 1,320 vials or 6,216 straws, making it an essential unit in scientific environments demanding durability and performance.