Learn More
System Biosciences LLC XCF EXOSOMAL DNA ISOLATION KITXCF Exosomal DNA Isolation Kit = 20 Reactions
SDP
Catalog No. NC1845132
Encompass_Preferred
Efficiently extract DNA from already-isolated exosomes with the XCF Exosomal DNA Isolation Kit. Delivering exosomal DNA ready for sequencing or other applications, the XCF™ Exosomal DNA Isolation Kit simplifies extraction of DNA from already isolated exosomes. Compatible with exosomes isolated using any of SBI’s ExoQuick products, ultracentrifugation, or other exosome isolation methods, the XCF Exosomal DNA Isolation Kit expands what you can learn from exosomes.
Catalog No. NC1845132 Supplier System Biosciences LLC Supplier No. XCF200A1
Efficiently extract DNA from already-isolated exosomes with the XCF Exosomal DNA Isolation Kit. Delivering exosomal DNA ready for sequencing or other applications, the XCF™ Exosomal DNA Isolation Kit simplifies extraction of DNA from already isolated exosomes. Compatible with exosomes isolated using any of SBI’s ExoQuick products, ultracentrifugation, or other exosome isolation methods, the XCF Exosomal DNA Isolation Kit expands what you can learn from exosomes.