Learn More
MSC Excel Dryer Inc. XChanger Combo Kit, ADA, Stainless Steel, Hand Dryer Retrofit
Catalog No. 502838484
Encompass
The XChanger Combo Kit retrofits a paper dispenser and waste receptacle with an XLERATOR or XLERATOReco hand dryer. It installs easily over the former dispenser's space without wall damage. An ADA height compliant version ensures full accessibility.
  • Allows complete retrofit of paper towel dispensers and waste receptacles with hand dryers.
  • Installs easily over the space left by former paper dispensers.
  • Prevents damage to the existing wall surface during installation.
  • Contributes to a modern, clean, and green restroom environment.
  • Available in an ADA height compliant version for accessibility.
  • Designed to minimize or eliminate the need for wall repair.
  • Recess kit adheres to 4" protrusion ADA guidelines.
  • Helps to keep water off the wall and floor.
Catalog No. 50-283-8484 Supplier MSC Supplier No. 38863288
The XChanger Combo Kit retrofits a paper dispenser and waste receptacle with an XLERATOR or XLERATOReco hand dryer. It installs easily over the former dispenser's space without wall damage. An ADA height compliant version ensures full accessibility.
  • Allows complete retrofit of paper towel dispensers and waste receptacles with hand dryers.
  • Installs easily over the space left by former paper dispensers.
  • Prevents damage to the existing wall surface during installation.
  • Contributes to a modern, clean, and green restroom environment.
  • Available in an ADA height compliant version for accessibility.
  • Designed to minimize or eliminate the need for wall repair.
  • Recess kit adheres to 4" protrusion ADA guidelines.
  • Helps to keep water off the wall and floor.