Learn More
Eurofins Discoverx Recombinant Human XCL1/Lymphotactin, 25 µg. (92-1092)
Supplier:  Eurofins Discoverx 921092
Encompass
XCL1 (lymphotactin) is a chemokine protein that functions in inflammation and immunological responses. Specifically chemotactic for T cells (lymphocytes), but not for monocytes or neutrophils. Binds and acts as an agonist to the chemokine receptor XCR1. Provided as 25 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9630