Learn More
New England Biolabs, Inc. XcmI – 5000 units
SDP
Catalog No. 50812197
Encompass
A restriction endonuclease that recognizes the sequence CCANNNN_N^NNNNTGG.
Catalog No. 50-812-197 Supplier New England Biolabs, Inc. Supplier No. R0533L
A restriction endonuclease that recognizes the sequence CCANNNN_N^NNNNTGG.