Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMOMETEC XcytoMatic 40 Consumable Pack
SDP

Catalog No. NC3811870
Encompass

Included in the XM40 consumables starter pack:

Reagents:
910-0017 - XM Rinse 1 – QTY 2
910-0018 - XM Cell Stain 2 – QTY 1
910-0019 - Cleaning solution – QTY 1

Labware:
911-0016 - Liquid waste 1, 2000 m – QTY 2
911-0017 - Tube waste 1, 4000 Ml – QTY 2
911-0018 - Sample tube 1 – QTY 1

Catalog No. NC3811870 Supplier CHEMOMETEC Supplier No. 9720041
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Included in the XM40 consumables starter pack:

Reagents:
910-0017 - XM Rinse 1 – QTY 2
910-0018 - XM Cell Stain 2 – QTY 1
910-0019 - Cleaning solution – QTY 1

Labware:
911-0016 - Liquid waste 1, 2000 m – QTY 2
911-0017 - Tube waste 1, 4000 Ml – QTY 2
911-0018 - Sample tube 1 – QTY 1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.