Learn More
Sklar Instruments Sklarlite XD Mayo-Hegar Needle Holder, Sklar, 7 in., Straight, Serrated, Extra Light and Extra Delicate (SklarLite XD), Finger Rings, Stainless Steel. EA/1.
SDP
Catalog No. 501437971 Shop All Sklar Instruments Products
Encompass_Preferred
Sklarlite XD Mayo-Hegar Needle Holder, Sklar, 7 in., Straight, Serrated, Extra Light and Extra Delicate (SklarLite XD), Finger Rings, Stainless Steel. The Sklarlite XD Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. This product is straight with serrated jaws, and measures 7 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7971 Supplier Sklar Instruments Supplier No. 232228
Sklarlite XD Mayo-Hegar Needle Holder, Sklar, 7 in., Straight, Serrated, Extra Light and Extra Delicate (SklarLite XD), Finger Rings, Stainless Steel. The Sklarlite XD Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths, and used in multiple types of surgical procedures. Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. This product is straight with serrated jaws, and measures 7 inches.