Learn More
MARSHALL SCIENTIFIC Edwards XDS46i Dry Scroll Pump
SDP
Catalog No. NC2888782
Encompass_Preferred
The XDS dry scroll pumps have become industry standard when dry pumping is essential, proving to be a robust and clean vacuum pump solution in a range of applications and processes. The XDS46 series shares many of the same features of the XDS35 series but has been optimized for maximum pumping speed at inlet pressures between 1 mbar and 10 mbar, making it ideally suited for LCMS and ICPMS applications requiring higher pumping speeds. Ultimate vacuum of 0.050 mbar and peak pumping speed of 23.5 CFM.
Catalog No. NC2888782 Supplier MARSHALL SCIENTIFIC Supplier No. EDA73101983
Add to Cart
Add to Cart
Request a Quote
The XDS dry scroll pumps have become industry standard when dry pumping is essential, proving to be a robust and clean vacuum pump solution in a range of applications and processes. The XDS46 series shares many of the same features of the XDS35 series but has been optimized for maximum pumping speed at inlet pressures between 1 mbar and 10 mbar, making it ideally suited for LCMS and ICPMS applications requiring higher pumping speeds. Ultimate vacuum of 0.050 mbar and peak pumping speed of 23.5 CFM.