Learn More
Medchemexpress LLC HY-108577 5mg Medchemexpress, XE 991 dihydrochloride CAS:122955-13-9 Purity:>98%
SDP
Catalog No. 502080747
Encompass_Preferred
Medchemexpress, HY-108577 5mg XE 991 dihydrochloride CAS:122955-13-9 XE 991 dihydrochloride, a Kv7 (KCNQ) channels blocker, potently inhibits Kv7.1 (KCNQ1), Kv7.2 (KCNQ2), Kv7.2 + Kv7.3 (KCNQ3) channel, and M-current with IC50s of 0.75 µM, 0.71 µM, 0.6 μM, and 0.98 µM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0747 Supplier Medchemexpress LLC Supplier No. HY1085775MG
Medchemexpress, HY-108577 5mg XE 991 dihydrochloride CAS:122955-13-9 XE 991 dihydrochloride, a Kv7 (KCNQ) channels blocker, potently inhibits Kv7.1 (KCNQ1), Kv7.2 (KCNQ2), Kv7.2 + Kv7.3 (KCNQ3) channel, and M-current with IC50s of 0.75 µM, 0.71 µM, 0.6 μM, and 0.98 µM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.