Learn More
Cayman Chemical XE 991 (hydrochloride), Purity: ≥98%, CAS: 122955-13-9, 5 mg
SDP
Catalog No. 502973189
Encompass_Preferred
A blocker of KCNQ channels that potently inhibits KCNQ1 and 2 homomeric channels (IC50 = 0.75 and 0.71 µM, respectively) as well as KCNQ2+3 heteromultimers (IC50 = 0.6 µM); shows good in vivo potency and duration of action
Catalog No. 50-297-3189 Supplier Cayman Chemical Supplier No. 145825 MG
A blocker of KCNQ channels that potently inhibits KCNQ1 and 2 homomeric channels (IC50 = 0.75 and 0.71 µM, respectively) as well as KCNQ2+3 heteromultimers (IC50 = 0.6 µM); shows good in vivo potency and duration of action