Learn More
Medchemexpress LLC XE991 dihydrochloride | 122955-13-9 | 99.6% | 449.37 | 10 MG
Supplier: Medchemexpress LLC HY10857750MG
XE991 dihydrochloride is a Kv7 (KCNQ) channel blocker that potently inhibits Kv7.1 (KCNQ1), Kv7.2 (KCNQ2), Kv7.2 + Kv7.3 (KCNQ3) channels, and M-current.
- Potently inhibits Kv7.1 (KCNQ1) channels with an IC50 of 0.75 μM.
- Inhibits Kv7.2 (KCNQ2) channels with an IC50 of 0.71 μM.
- Blocks Kv7.2 + Kv7.3 (KCNQ3) channels with an IC50 of 0.6 μM.
- Inhibits M-current with an IC50 of 0.98 μM.
- Enhances [3H]ACh release from rat brain slices with an EC50 of 490 nM.
- Demonstrates good in vivo potency and duration of action.