Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC XE991 dihydrochloride | 122955-13-9 | 99.6% | 449.37 | 500 UG
SDP

Catalog No. 5000345273
Encompass_Preferred

XE991 dihydrochloride is a Kv7 (KCNQ) channels blocker that potently inhibits Kv7.1 (KCNQ1), Kv7.2 (KCNQ2), Kv7.2 + Kv7.3 (KCNQ3) channel, and M-current with IC50s of 0.75 μM, 0.71 μM, 0.6 μM, and 0.98 μM, respectively. It has an EC50 of 490 nM for enhancement of [3H]ACh release from rat brain slices, and shows good in vivo potency and duration of action.

  • Potent Kv7 (KCNQ) channels blocker.
  • Inhibits Kv7.1, Kv7.2, Kv7.2 + Kv7.3 channels, and M-current.
  • IC50s: 0.75 μM (Kv7.1), 0.71 μM (Kv7.2), 0.6 μM (Kv7.2 + Kv7.3), 0.98 μM (M-current).
  • EC50 of 490 nM for enhancing [3H]ACh release from rat brain slices.
  • Good in vivo potency and duration of action.

Catalog No. 50-003-45273 Supplier Medchemexpress LLC Supplier No. HY10857710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XE991 dihydrochloride is a Kv7 (KCNQ) channels blocker that potently inhibits Kv7.1 (KCNQ1), Kv7.2 (KCNQ2), Kv7.2 + Kv7.3 (KCNQ3) channel, and M-current with IC50s of 0.75 μM, 0.71 μM, 0.6 μM, and 0.98 μM, respectively. It has an EC50 of 490 nM for enhancement of [3H]ACh release from rat brain slices, and shows good in vivo potency and duration of action.

  • Potent Kv7 (KCNQ) channels blocker.
  • Inhibits Kv7.1, Kv7.2, Kv7.2 + Kv7.3 channels, and M-current.
  • IC50s: 0.75 μM (Kv7.1), 0.71 μM (Kv7.2), 0.6 μM (Kv7.2 + Kv7.3), 0.98 μM (M-current).
  • EC50 of 490 nM for enhancing [3H]ACh release from rat brain slices.
  • Good in vivo potency and duration of action.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.