Learn More
Selleck Chemical LLC XE991 dihydrochloride S2967-25mg
SDP
Catalog No. 503111617
Encompass_Preferred
XE 991 (LS190926) dihydrochloride is a blocker of Kv7 (KCNQ) channels that potently inhibits KCNQ1 (Kv7 1) KCNQ2 (Kv7 2) KCNQ2 KCNQ3 (Kv7 3) channel and M-current with IC50 of 0 75 M 0 71 M 0 6 M and 0 98 M respectively
Catalog No. 50-311-1617 Supplier Selleck Chemical LLC Supplier No. S296725MG
Add to Cart
Add to Cart
XE 991 (LS190926) dihydrochloride is a blocker of Kv7 (KCNQ) channels that potently inhibits KCNQ1 (Kv7 1) KCNQ2 (Kv7 2) KCNQ2 KCNQ3 (Kv7 3) channel and M-current with IC50 of 0 75 M 0 71 M 0 6 M and 0 98 M respectively