Learn More
Labelmaster Inc Dustless Polypropylene Zip-Lock Pouch for 1.5 oz. Vial
SDP
Catalog No. NC9064520
Encompass_Preferred
This Sorbent Lined Pouch for Vials is great for packaging liquid products. The pouch includes absorbent and poly bag. Our easy-to-use, zip-lock 3 in. x 5 in. pouch is filled with polypropylene absorbent, which will absorb up to 1.5 oz.
Catalog No. NC9064520 Supplier Labelmaster Inc Supplier No. KSXP15R1
This Sorbent Lined Pouch for Vials is great for packaging liquid products. The pouch includes absorbent and poly bag. Our easy-to-use, zip-lock 3 in. x 5 in. pouch is filled with polypropylene absorbent, which will absorb up to 1.5 oz.