Learn More
Bachem Xenin 25
SDP
Supplier:  Bachem 4031355.0500
Encompass_Preferred
Xenin 25, H-Met-Leu-Thr-Lys-Phe-Glu-Thr-Lys-Ser-Ala-Arg-Val-Lys-Gly-Leu-Ser-Phe-His-Pro-Lys-Arg-Pro-Trp-Ile-Leu-OH, 144092-28-4, C139H224N38O32S, Mr 2971.61, 0.5mg. Xenin. Xenin 25, which has 6 C-terminal amino acids in common with amphibian xenopsin, has been detected in human gastric mucosa. It stimulates the exocrine pancreatic secretion. Xenin co-excreted with the incretin GIP has gained interest in diabetes research.
Catalog No. 50-194-6913