Learn More
MSC Miller Xenon Automatic Pass-Through Shuttle, Stainless Steel, 310 lb Capacity, 1 ea
Catalog No. 5000509788
Encompass
The Xenon Automatic Pass-Through Shuttle is designed for use with permanent horizontal lifeline systems to enhance worker safety and mobility. This shuttle features an automatic pass-through design that allows users to navigate across intermediate supports without the need to disconnect from the lifeline, ensuring continuous protection during overhead applications.
  • Automatic pass-through design for smooth, hands-free navigation across intermediate supports
  • Double-locking mechanism enables simple, one-handed operation
  • Stainless steel construction provides durability and corrosion resistance in harsh environments
  • Self-aligning attachment ring facilitates passage through brackets
  • Compatible with permanent horizontal lifeline systems
  • Rated for a maximum worker weight limit of 310 lb
Catalog No. 50-005-09788 Supplier MSC Supplier No. 59817924
The Xenon Automatic Pass-Through Shuttle is designed for use with permanent horizontal lifeline systems to enhance worker safety and mobility. This shuttle features an automatic pass-through design that allows users to navigate across intermediate supports without the need to disconnect from the lifeline, ensuring continuous protection during overhead applications.
  • Automatic pass-through design for smooth, hands-free navigation across intermediate supports
  • Double-locking mechanism enables simple, one-handed operation
  • Stainless steel construction provides durability and corrosion resistance in harsh environments
  • Self-aligning attachment ring facilitates passage through brackets
  • Compatible with permanent horizontal lifeline systems
  • Rated for a maximum worker weight limit of 310 lb