Learn More
Agilent Technologies Pulsed Xenon Flash Lamp, For Fluorescence Detectors, 1 ea
Catalog No. NC2385242
Encompass
This genuine pulsed xenon flash lamp is specifically designed as the excitation light source for fluorescence detectors in liquid chromatography systems. It is intended to restore or maintain the excitation output necessary for accurate fluorescence measurements, ensuring continued system sensitivity and reproducibility in analytical workflows.
  • Provides reliable excitation for compatible fluorescence detector modules.
  • Maintains consistent fluorescence sensitivity and measurement reproducibility.
  • Engineered to meet specific quality and traceability standards for seamless integration.
  • Supports direct compatibility with specified detector modules to simplify procurement and fitment.
  • Replacement is intended as an advanced maintenance procedure for improved safety and reliability.
Catalog No. NC2385242 Supplier Agilent Technologies Supplier No. 21400600
This genuine pulsed xenon flash lamp is specifically designed as the excitation light source for fluorescence detectors in liquid chromatography systems. It is intended to restore or maintain the excitation output necessary for accurate fluorescence measurements, ensuring continued system sensitivity and reproducibility in analytical workflows.
  • Provides reliable excitation for compatible fluorescence detector modules.
  • Maintains consistent fluorescence sensitivity and measurement reproducibility.
  • Engineered to meet specific quality and traceability standards for seamless integration.
  • Supports direct compatibility with specified detector modules to simplify procurement and fitment.
  • Replacement is intended as an advanced maintenance procedure for improved safety and reliability.