Learn More
GENERAL BIOPHYSICS LLC Xenon Gas Analyzer Model A
Catalog No. NC3361667
Encompass
Thermoconductivity Gas Analyzer for Xenon with Onboard PumpApplication: Calibrated for xenon gas with onboard pump to take gas samples.Features: Includes data display screen.Accuracy: ±0.3%.
Catalog No. NC3361667 Supplier GENERAL BIOPHYSICS LLC Supplier No. GAXEO2A
Add to Cart
Add to Cart
Request a Quote
Thermoconductivity Gas Analyzer for Xenon with Onboard PumpApplication: Calibrated for xenon gas with onboard pump to take gas samples.Features: Includes data display screen.Accuracy: ±0.3%.