Learn More
GENERAL BIOPHYSICS LLC Xenon Gas Analyzer Model B
Catalog No. NC3361668
Encompass
Thermoconductivity Gas Analyzer for Xenon (In-line Monitoring)Application: Real-time, in-line concentration monitoring for xenon gas.Accuracy: ±1%.
Catalog No. NC3361668 Supplier GENERAL BIOPHYSICS LLC Supplier No. GAXEO2B
Thermoconductivity Gas Analyzer for Xenon (In-line Monitoring)Application: Real-time, in-line concentration monitoring for xenon gas.Accuracy: ±1%.