Learn More
Analytical Sales & Services Xenon Lamp & Holder For Rainin
Catalog No. 5000806578
Encompass
Xenon Lamp & Holder For Rainin/Cary/Varian Model 9070 9075 FL-1 2 PRO STAR 360
Qty/Pkg: EACH
Catalog No. 50-008-06578 Supplier Analytical Sales & Services Supplier No. 128910
Xenon Lamp & Holder For Rainin/Cary/Varian Model 9070 9075 FL-1 2 PRO STAR 360
Qty/Pkg: EACH