Learn More
Perkin Elmer US LLC Xenon Pulse Lamp with Silencer
Catalog No. 502112301
Encompass_Preferred
Our Fluorescence Spectrometers use Pulsed Xenon Discharge lamps. These reliable, stable lamps provide remarkable sensitivity and can achieve a peak intensity exceeding 1 KW. For optimal instrument perfomance, always use original PerkinElmer replacement lamps.
Catalog No. 50-211-2301 Supplier Perkin Elmer US LLC Supplier No. N4202020
Our Fluorescence Spectrometers use Pulsed Xenon Discharge lamps. These reliable, stable lamps provide remarkable sensitivity and can achieve a peak intensity exceeding 1 KW. For optimal instrument perfomance, always use original PerkinElmer replacement lamps.