Learn More
Bulbman Inc XBOR100W/45C-OFR Osram 69191
SDP
Catalog No. NC9737762
Encompass_Preferred
Osram Sylvania 69191 XBO R 100 W/45C OFR 100 watt, xenon short arc, with reflector. Applications: Light guide, Baroscopic, Fiber Optic, Minimal Invasive Surgery, Endoscopy
Catalog No. NC9737762 Supplier Bulbman Inc Supplier No. 108874
Osram Sylvania 69191 XBO R 100 W/45C OFR 100 watt, xenon short arc, with reflector. Applications: Light guide, Baroscopic, Fiber Optic, Minimal Invasive Surgery, Endoscopy