Learn More
Moc Incorporated XENOPHOT HLX OSRAM 5/PK
Catalog No. NC9734808
Encompass
Lamp; Halogen; XENOPHOT;; Low-voltage lamps without reflector; Luminous flux (lm): 3600; Diameter d(mm): 11.5; Average life 50 hr.
Catalog No. NC9734808 Supplier Moc Incorporated Supplier No. 64625
Lamp; Halogen; XENOPHOT;; Low-voltage lamps without reflector; Luminous flux (lm): 3600; Diameter d(mm): 11.5; Average life 50 hr.