Learn More
Hach Company Xenosep extraction thimbles
Supplier:  Hach Company 2966600
Encompass
Soxhlet Thimbles-Cellulose-22x100mm-25/PKXenosep extraction thimbles are ideal for determining the concentration of nonvolatile and semivolatile organic compounds from environmental solid waste matrices sludge soil air and water The thimbles are manufactured from high purity alpha cellulose cotton linters which provide excellent mechanical strength stability and retention with very low/no background extractables in a disposable sample holder format
Catalog No. 50-002-07193