Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Xeruborbactam disodium | 2170848-99-2 | 99.0% | 283.96 | 5 MG
SDP

Catalog No. 5000364321
Encompass_Preferred

Xeruborbactam disodium | 2170848-99-2 | 99.0% | 283.96 | 5 MG

Catalog No. 50-003-64321 Supplier Medchemexpress LLC Supplier No. 5MGHY136072
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Xeruborbactam disodium | 2170848-99-2 | 99.0% | 283.96 | 5 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.