Learn More
Cayman Chemical XestospongIn C 100UG
SDP
Catalog No. 502896247
Encompass_Preferred
An inositol-1,4,5-trisphosphate receptor antagonist; completely blocks the increase in intracellular calcium in vascular smooth muscle cells at a concentration of 25 µM
Catalog No. 50-289-6247 Supplier Cayman Chemical Supplier No. 64950100 UG
An inositol-1,4,5-trisphosphate receptor antagonist; completely blocks the increase in intracellular calcium in vascular smooth muscle cells at a concentration of 25 µM