Learn More
Cayman Chemical XestospongIn C 10UG
SDP
Catalog No. 502892609
Encompass_Preferred
An inositol-1,4,5-trisphosphate receptor antagonist; completely blocks the increase in intracellular calcium in vascular smooth muscle cells at a concentration of 25 µM
Catalog No. 50-289-2609 Supplier Cayman Chemical Supplier No. 6495010 UG
An inositol-1,4,5-trisphosphate receptor antagonist; completely blocks the increase in intracellular calcium in vascular smooth muscle cells at a concentration of 25 µM