Learn More
Abcam x estospongin C, IP3 receptor antagonist, 10UG
Catalog No. 502584548
Encompass_Preferred
MW 446.72 g/mol, Purity >90%. Potent, selective, reversible IP₃ receptor antagonist (IC₅₀ = 350 nM). Inhibits IP₃-mediated Ca²+ release from ER and SR. 30-Fold selectivity over ryanodine receptor. Inhibits voltage-dependent Ca²+ and K+ channels at higher concentrations. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4548 Supplier Abcam Supplier No. AB12091410UG
MW 446.72 g/mol, Purity >90%. Potent, selective, reversible IP₃ receptor antagonist (IC₅₀ = 350 nM). Inhibits IP₃-mediated Ca²+ release from ER and SR. 30-Fold selectivity over ryanodine receptor. Inhibits voltage-dependent Ca²+ and K+ channels at higher concentrations. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement