Learn More
Waters Corp Performance Maintenance Kit, Xevo G2-S, LockSpray, MS System
Catalog No. 502391003 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the Xevo G2-S (LockSpray / rotary) Mass Spectrometer contains all the Waters Quality Parts™ necessary to keep the instrument operating at peak performance on an annual basis. This kit includes Source components, ESI probe components and vacuum pump components.
  • Kit contains all necessary parts to maintain instrument operating at peak performance.
  • Designed for an annual replacement schedule.
  • Includes source components.
  • Includes ESI probe components.
  • Includes vacuum pump components.
  • Compatible with MS system types.
  • Units per package: 1.
Catalog No. 50-239-1003 Supplier Waters Corp Supplier No. 201000276
The Waters™ Performance Maintenance Kit for the Xevo G2-S (LockSpray / rotary) Mass Spectrometer contains all the Waters Quality Parts™ necessary to keep the instrument operating at peak performance on an annual basis. This kit includes Source components, ESI probe components and vacuum pump components.
  • Kit contains all necessary parts to maintain instrument operating at peak performance.
  • Designed for an annual replacement schedule.
  • Includes source components.
  • Includes ESI probe components.
  • Includes vacuum pump components.
  • Compatible with MS system types.
  • Units per package: 1.