Learn More
Waters Corp NanoLockSpray Exact Mass Ionization Source, Xevo G2-S Compatible
Catalog No. 502392099 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This NanoLockSpray dual electrospray ion source provides authenticated exact mass measurement in NanoFlow LC-MS and LC-MS/MS modes. It optimizes co-introduction of analyte and lock mass compounds directly into the ion source, offering superior exact mass data acquisition. This increases specificity for compound or fragment identification in metabolite profiling and proteomics.
  • Provides authenticated exact mass measurement in NanoFlow LC-MS and LC-MS/MS modes.
  • Features optimized co-introduction of analyte and lock mass compounds directly into the ion source.
  • Enhances compound or fragment identification in metabolite profiling and proteomics.
  • Overcomes limitations of traditional methods, such as ionization suppression and mass interferences.
  • Utilizes a dual electrospray system for lock mass reference introduction.
  • Compatible with Xevo G2-S Tof and Xevo G2-S QTof systems.
Catalog No. 50-239-2099 Supplier Waters Corp Supplier No. 176003137
This NanoLockSpray dual electrospray ion source provides authenticated exact mass measurement in NanoFlow LC-MS and LC-MS/MS modes. It optimizes co-introduction of analyte and lock mass compounds directly into the ion source, offering superior exact mass data acquisition. This increases specificity for compound or fragment identification in metabolite profiling and proteomics.
  • Provides authenticated exact mass measurement in NanoFlow LC-MS and LC-MS/MS modes.
  • Features optimized co-introduction of analyte and lock mass compounds directly into the ion source.
  • Enhances compound or fragment identification in metabolite profiling and proteomics.
  • Overcomes limitations of traditional methods, such as ionization suppression and mass interferences.
  • Utilizes a dual electrospray system for lock mass reference introduction.
  • Compatible with Xevo G2-S Tof and Xevo G2-S QTof systems.