Learn More
Waters Corp Service Kit, Xevo TQ-GC, CI Source, Annual Replacement
Catalog No. 502391001 Shop All Waters Corporation Products
Encompass_Preferred
This service kit is designed for maintaining a CI Source on a Xevo TQ-GC Mass Spectrometer. It includes essential replacement components to ensure continued functionality, ensuring optimal performance and longevity of your equipment.
  • Includes filament assembly.
  • Includes CI ion chamber.
  • Designed for mass spectrometer systems.
  • Recommended for annual replacement.
  • A rebuild kit for maintaining your system.
Catalog No. 50-239-1001 Supplier Waters Corp Supplier No. 700012077
This service kit is designed for maintaining a CI Source on a Xevo TQ-GC Mass Spectrometer. It includes essential replacement components to ensure continued functionality, ensuring optimal performance and longevity of your equipment.
  • Includes filament assembly.
  • Includes CI ion chamber.
  • Designed for mass spectrometer systems.
  • Recommended for annual replacement.
  • A rebuild kit for maintaining your system.