Learn More
Waters Corp Xevo TQ-GC Performance Maintenance Kit, MS System, Annual, Chemical Kit
Catalog No. 502392837 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the Xevo TQ-GC Mass Spectrometer contains all the Waters Quality Parts™ necessary to keep the instrument operating at peak performance. This particular kit also includes a chemical kit which is required to complete the performance testing.
  • Includes high-quality replacement parts.
  • Includes source components.
  • Includes vacuum pump components.
  • Delivers a chemical kit for performance testing.
Catalog No. 50-239-2837 Supplier Waters Corp Supplier No. 176004385
Add to Cart
Add to Cart
Request a Quote
The Waters™ Performance Maintenance Kit for the Xevo TQ-GC Mass Spectrometer contains all the Waters Quality Parts™ necessary to keep the instrument operating at peak performance. This particular kit also includes a chemical kit which is required to complete the performance testing.
  • Includes high-quality replacement parts.
  • Includes source components.
  • Includes vacuum pump components.
  • Delivers a chemical kit for performance testing.