Learn More
Waters Corp Performance Maintenance Kit, Xevo TQ-GC, CI Service, Chemical Kit
Catalog No. 502391727 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Performance Maintenance Kit for the Xevo TQ-GC Mass Spectrometer contains all the quality parts necessary to keep the instrument operating at peak performance. It includes source components and vacuum pump components. This particular part number also delivers a chemical kit, which is required to complete the performance testing. Additionally, an extra service kit for the CI optional source is provided.
  • Contains all parts necessary to keep instrument operating at peak performance.
  • Includes source components and vacuum pump components.
  • Delivers a chemical kit required for performance testing.
  • Provides an extra service kit for the CI optional source.
Catalog No. 50-239-1727 Supplier Waters Corp Supplier No. 176004386
Add to Cart
Add to Cart
Request a Quote
The Performance Maintenance Kit for the Xevo TQ-GC Mass Spectrometer contains all the quality parts necessary to keep the instrument operating at peak performance. It includes source components and vacuum pump components. This particular part number also delivers a chemical kit, which is required to complete the performance testing. Additionally, an extra service kit for the CI optional source is provided.
  • Contains all parts necessary to keep instrument operating at peak performance.
  • Includes source components and vacuum pump components.
  • Delivers a chemical kit required for performance testing.
  • Provides an extra service kit for the CI optional source.