Learn More
Waters Corp WATERS CORPORATION Standards, Mass Spectrometry, Xevo TQ-S, Liquid, Room Temperature
Catalog No. 50201694 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product provides Mass Spectrometry (Xevo TQ-S) standards for system assessment, calibration, and benchmarking performance. It ensures quality results and comes with a Certificate of Analysis.
  • For Mass Spectrometry system assessment, calibration, and performance benchmarking
  • Quality assurance with Certificate of Analysis
  • Two components: calibration solution and resolution solution
  • Calibration solution: 1 x 30 mL Ortho-Phosphoric Acid 0.1% Acetonitrile/Water (1:1 V/V)
  • Resolution solution: 1 x 30 mL with Sodium Iodide 2 μg/ul, Cesium Iodide 50 ng/ul, Methyl Sulfate Sodium Salt 20 ng/μl in IPA/Water (1:1 V/V)
  • Liquid formulation, room temperature storage
  • 12 months shelf life
  • 30 mL volume
  • Hazardous material, special freight handling required
Catalog No. 50-201-694 Supplier Waters Corp Supplier No. 700005471
This product provides Mass Spectrometry (Xevo TQ-S) standards for system assessment, calibration, and benchmarking performance. It ensures quality results and comes with a Certificate of Analysis.
  • For Mass Spectrometry system assessment, calibration, and performance benchmarking
  • Quality assurance with Certificate of Analysis
  • Two components: calibration solution and resolution solution
  • Calibration solution: 1 x 30 mL Ortho-Phosphoric Acid 0.1% Acetonitrile/Water (1:1 V/V)
  • Resolution solution: 1 x 30 mL with Sodium Iodide 2 μg/ul, Cesium Iodide 50 ng/ul, Methyl Sulfate Sodium Salt 20 ng/μl in IPA/Water (1:1 V/V)
  • Liquid formulation, room temperature storage
  • 12 months shelf life
  • 30 mL volume
  • Hazardous material, special freight handling required