Learn More
Waters Corp Performance Maintenance Kit, Xevo TQ-S cronos (Rotary), MS, Annual
Catalog No. 502391795 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Performance Maintenance Kit is designed for the Xevo TQ-S cronos (rotary) Mass Spectrometer. It contains all necessary quality parts to ensure the instrument operates at peak performance on an annual basis. The kit includes essential source components and vacuum pump components, though a separate Probe Assembly is recommended for optimal performance.
  • Product type: Performance maintenance kits
  • System type: MS
  • Replacement schedule: Annual
  • Components included: Source components and vacuum pump components
Catalog No. 50-239-1795 Supplier Waters Corp Supplier No. 201000322
This Performance Maintenance Kit is designed for the Xevo TQ-S cronos (rotary) Mass Spectrometer. It contains all necessary quality parts to ensure the instrument operates at peak performance on an annual basis. The kit includes essential source components and vacuum pump components, though a separate Probe Assembly is recommended for optimal performance.
  • Product type: Performance maintenance kits
  • System type: MS
  • Replacement schedule: Annual
  • Components included: Source components and vacuum pump components