Learn More
Waters Corp Installation Standards Kit, for Xevo TQ-S micro System
Catalog No. 502390562 Shop All Waters Corporation Products
Encompass_Preferred
This kit is essential for the correct installation and setup of the Xevo TQ-S micro System mass spectrometer in a laboratory. It contains various solutions and compounds vital for ensuring sensitive analysis, large dynamic range, and high data acquisition rates, leading to accurate and repeatable findings.
  • Essential for mass spectrometer system installation.
  • Ensures correct laboratory setup.
  • Includes set up solution (25 mL) and stock solution (25 mL).
  • Contains chloramphenicol (50 mg), sulfadimethoxine (5 mg), leucine enkephalin (5 mg), aqueous ammonia (3 mL), and formic acid (1 mL).
  • Facilitates detection of low-concentration analytes in complex matrices using ZSpray™ and StepWave™ technologies.
  • Enables low volume injections for accurate, repeatable results.
  • Hazardous material requiring special freight handling.
Catalog No. 50-239-0562 Supplier Waters Corp Supplier No. 186007591
This kit is essential for the correct installation and setup of the Xevo TQ-S micro System mass spectrometer in a laboratory. It contains various solutions and compounds vital for ensuring sensitive analysis, large dynamic range, and high data acquisition rates, leading to accurate and repeatable findings.
  • Essential for mass spectrometer system installation.
  • Ensures correct laboratory setup.
  • Includes set up solution (25 mL) and stock solution (25 mL).
  • Contains chloramphenicol (50 mg), sulfadimethoxine (5 mg), leucine enkephalin (5 mg), aqueous ammonia (3 mL), and formic acid (1 mL).
  • Facilitates detection of low-concentration analytes in complex matrices using ZSpray™ and StepWave™ technologies.
  • Enables low volume injections for accurate, repeatable results.
  • Hazardous material requiring special freight handling.