Learn More
Waters Corp Performance Maintenance Kit, Xevo TQ-S micro (Rotary), Annual, Chemical Kit
Catalog No. 502392196 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This performance maintenance kit is designed for the Xevo TQ-S micro (rotary) Mass Spectrometer. It contains all the necessary quality parts to ensure the instrument operates at peak performance on an annual basis and includes a chemical kit required for performance testing.
  • Designed for MS system type
  • Annual replacement schedule
  • Includes source components
  • Includes ESI probe components
  • Includes vacuum pump components
  • Comes with a chemical kit for performance testing
  • One unit per package
Catalog No. 50-239-2196 Supplier Waters Corp Supplier No. 176003850
This performance maintenance kit is designed for the Xevo TQ-S micro (rotary) Mass Spectrometer. It contains all the necessary quality parts to ensure the instrument operates at peak performance on an annual basis and includes a chemical kit required for performance testing.
  • Designed for MS system type
  • Annual replacement schedule
  • Includes source components
  • Includes ESI probe components
  • Includes vacuum pump components
  • Comes with a chemical kit for performance testing
  • One unit per package