Learn More
Waters Corp Performance Maintenance Kit, Xevo TQ-S Micro Mass Spectrometer, Annual
Catalog No. 502391542 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Performance Maintenance Kit for the Xevo TQ-S micro (rotary) Mass Spectrometer contains all necessary quality parts to keep your instrument operating at peak performance on an annual basis. It includes source components, ESI probe components, and vacuum pump components.
  • Includes source components.
  • Includes ESI probe components.
  • Includes vacuum pump components.
  • Designed for Xevo TQ-S micro Mass Spectrometer.
  • Annual replacement schedule.
  • One unit per package.
Catalog No. 50-239-1542 Supplier Waters Corp Supplier No. 201000294
The Performance Maintenance Kit for the Xevo TQ-S micro (rotary) Mass Spectrometer contains all necessary quality parts to keep your instrument operating at peak performance on an annual basis. It includes source components, ESI probe components, and vacuum pump components.
  • Includes source components.
  • Includes ESI probe components.
  • Includes vacuum pump components.
  • Designed for Xevo TQ-S micro Mass Spectrometer.
  • Annual replacement schedule.
  • One unit per package.