Learn More
Waters Corp Performance Maintenance Kit, Xevo TQ-S micro, Scroll, Chemical Kit
Catalog No. 502391848 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the Xevo TQ-S micro (scroll) Mass Spectrometer contains all the Waters Quality Parts™ necessary to keep the instrument operating at peak performance on an annual basis. This particular kit also includes a chemical kit required for performance testing.
  • Contains source, ESI probe, and vacuum pump components.
  • Designed for annual replacement.
  • Includes a chemical kit for performance testing.
  • One kit per package.
Catalog No. 50-239-1848 Supplier Waters Corp Supplier No. 176003851
The Waters™ Performance Maintenance Kit for the Xevo TQ-S micro (scroll) Mass Spectrometer contains all the Waters Quality Parts™ necessary to keep the instrument operating at peak performance on an annual basis. This particular kit also includes a chemical kit required for performance testing.
  • Contains source, ESI probe, and vacuum pump components.
  • Designed for annual replacement.
  • Includes a chemical kit for performance testing.
  • One kit per package.